Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
Click to view available options
Diametro (sistema metrico):
11.5 mm
29 mm
34 mm
Lunghezza (sistema metrico):
1.1 mL/cm vol./length
5 m
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Quantità:
1 m Tubing/Kit
5 m Tubing/Kit
Dimensione della confezione:
1m
5m
Pezzo
Descrizione
Includes
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifica
Specifica
| MWCO | 3.5 kDa |
| Diametro (sistema metrico) | 34 mm |
| Lunghezza (sistema metrico) | 9.3 mL/cm vol./length |
| Quantità | 1 m Tubing/Kit |