Learn More
Cellpath Stainless-steel Reusable Base Molds
Click to view available options
Dimensioni (L x P x A):
7 x 7 x 5 mm
10 x 24 x 5 mm
14 x 24 x 5 mm
24 x 24 x 5 mm
30 x 24 x 9 mm
37 x 24 x 9 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Made of reusable Stainless steel
- Stackable to reduce storage area
- Internal rounded corners enable easy removal of the wax block
- Features extended edges prevent tipping and provide grip area for ease of handling
- Available in wide range of sizes
- GBB-0714-05A suitable for use with needle and punch biopsies
Specifica
Specifica
| Materiale | Stainless Steel |
| Dimensioni (L x P x A) | 7 x 7 x 5 mm |
| Quantità | 5/Pk. |
Titolo del prodotto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido