Learn More
Solder, Lead-Free, 50g, ARMI∣MBH
Codice prodotto. 17970836
Click to view available options
Quantità:
50 g
Dimensione della confezione:
50g
Codice prodotto. 17970836
Marca: ARMI/MBH MBH74X CA2 CC
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Solder, Lead-Free, 50g
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Aluminium/Gold/Selenium/Zinc/Cobalt/Iron/Mercury/Phosphorus/Cadmium/Indium/Germanium/Arsenic/Bismuth/Nickel/Lead/Antimony/Copper/Silver
CAS 7440-31-5
Quantità 50 g
Forma Pieces
Concentration or Composition (by Analyte or Components) Aluminium (0.001) %/Gold (0.001) %/Selenium (0.001) %/Zinc (0.002) %/Cobalt (0.003) %/Iron (0.003) %/Mercury (0.003) %/Phosphorus (0.003) %/Cadmium (0.004) %/Indium (0.007) %/Germanium (0.01) %/Arsenic (0.015) %/Bismuth (0.031) %/Nickel (0.033) %/Lead (0.055) %/Antimony (0.08) %/Copper (0.8) %/Silver (3.5) %