Learn More
DiscoverX Recombinant Human Neuregulin-1 (Heregulin β2)
Codice prodotto. 17322429
Encompass
Click to view available options
:
500 μg
Codice prodotto. 17322429
Marca: DiscoverX 921091
Recombinant Human Neuregulin-1 (Heregulin β2) is an agonist for ErbB3 and ErbB4. Provided as 50μg of dry powder.
Neuregulin-1 is a glycoprotein that acts on EGFR family receptors, binding to and activating ErbB3 and ErbB4. Plays a critical role in growth and development, heart structure and function, and cardiomyocyte differentiation. Involved in development and repair of the peripheral nervous system. Provided as 50μg of dry powder.
Specifica
Quantità 50 μg
Product Type Control Ligands