Learn More
Hamilton™ 600 Microliter Syringes, No Needle Included
Codice prodotto. p-7121963
Click to view available options
Volume (sistema metrico):
2.5 μL
5 μL
Dimensione della confezione:
Pezzo
Codice prodotto. 11522505
Marca: Hamilton™ 763201
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0.25 μL up to 5 μL. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.
  • Half stroke version of the original icroliter 700 series syringes
  • Ideal for dispensing volumes from 0.25 μL up to 5 μL
  • Often used in applications that require one handed operation because of the short stroke length and the reinforced plunger
Specifica
Calibro dell'ago 22s G
Volume (sistema metrico) 2.5 μL