Learn More
YSI™ 6051030 Pro Series ISE/Conductivity Cable
Click to view available options
Lunghezza (sistema imperiale):
393.7 in.
Dimensione della confezione:
Pezzo
Descrizione
Measures any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), conductivity, salinity, specific conductance, total dissolved solids and temperature
- Conductivity sensor is built into the cable
- Cables over 1-meter also come with the 603062 cable management kit
- The 6051030 cables only work with the YSI Pro Plus handheld
- Does not include ISE sensor 15-176-209 (order separately)
Specifica
Specifica
| Da utilizzare con (applicazione) | Conductivity, pH, Temp |
| Include | Conductivity sensor |
| Lunghezza (sistema imperiale) | 393.7 in. |
| Lunghezza (sistema metrico) | 10 m |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur