Learn More
YSI™ 60520 Pro Series DO Cable
Descrizione
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty.Measure pH quickly and easily with the 1001 sensor.The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time information.
Cables over 1-meter also come with the 603062 cable management kit.
Order Info
Cables do NOT come with the sensor
Specifica
Specifica
| Tipo | Cable Assembly |
| Da utilizzare con (applicazione) | Measure temperature and DO |
| Lunghezza (sistema imperiale) cavo | 328 ft. |
| Lunghezza (sistema metrico) cavo | 100 m |
| Warranty | 2 Years |