Learn More
Integra Biosciences™ 8-Channel Aspiration Adapter
Click to view available options
Tipo:
8-channel Adapter
Dimensione della confezione:
Pezzo
Descrizione
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
- All parts in contact with fluids are autoclavable
Specifica
Specifica
| Tipo | 8-channel Adapter |
| Include | with ejector |