Learn More
Thermo Scientific™ 96-Well Flipper™ Microtube Racks
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Descrizione
Protect samples and allow racks to be stacked using the Thermo Scientific™ 96-Well Flipper™ Microtube Rack. The rack, which is molded with an alphanumeric grid for easy tube identification, holds 0.5mL tubes on one side and 1.5mL tubes on the other.
- Holds 0.5mL tubes on one side, 1.5mL tubes on the other
- Molded with an alphanumeric grid for easy tube identification
- Several colors available
Specifica
Specifica
| Descrizione | Rack flipper |
| Colore | Rosa fluorescente |
| Capacità | Microtubes (2 mL), Microtubes (1.5 mL), Microtubes (2 mL) |
| Materiale | Polipropilene |
| Forma | Rettangolare |
| Sovrapponibile | Sì |
| N. per custodia | 10 rack |
| Product Type | 96-well Flipper Rack |