Learn More
Honeywell Safety Products™ A800 Grey TSR Glasses
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Impact-resistant
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
- General use and protection against impact (45m/s).
- Well suited for indoor or outdoor applications such as utilities, light and heavy manufacturing, bio tech, laboratories, clean rooms, ship building and automotive manufacturing.
- Affordably priced and economical.
- Slim profile for workers with smaller faces.
- Sporty styling.
- 9-base polycarbonate, abrasion resistant lenses.
- 180° field of distortion-free vision.
- Temples feature soft, padded inserts with extra flex.
- Soft nosepiece conforms to various nose bridge shapes.
Specifica
Specifica
| Certificazione/Conformità | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1790, EC Attestation Number EC1790 |
| Lens Coating | Anti-fog, Impact-resistant |
| Lens Tint | Gray |
| Product Type | Safety Glasses |
| Da utilizzare con (applicazione) | Administration. General use and protection against impact (45m/s). Well suited for indoor/outdoor applications such as utilities, light and heavy manufacturing, bio tech, laboratories, clean rooms, ship building and automotive manufacturing. |
| Colore telaio | Gray |
| N. per confezione | 1 ciasc. |
| Lens Material | Policarbonato |