missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD3 Mouse anti-Human, Clone: 1G7G5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66697-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum"" disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.Specifica
| ABCD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28288 | |
| ABCD3 | |
| ABCD3 Fusion Protein Ag24760 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| Western Blot | |
| 1G7G5 | |
| Unconjugated | |
| ABCD3 | |
| ABC43, ABCD3, PMP70, PXMP1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5825 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto