missing translation for 'onlineSavingsMsg'
Learn More

AC260584, MedChemExpress

Codice prodotto. p-200100891
Encompass_Preferred
Click to view available options
Quantità:
1 mg
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
10mg
1mg
50mg
5mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16482948

Marca: Medchem Express HY100336

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress AC260584 is an M1 muscarinic receptor allosteric agonist with a pEC50 of 7.6.

AC260584 is an M1 muscarinic receptor allosteric agonist with a pEC50 of 7.6.

Specifications

Materiale o nome chimico AC260584
CAS 560083-42-3
Colore Light Yellow
Note sul grado di purezza Research
Formula molecolare C20H29FN2O2
Quantità 10 mg
Informazioni di solubilità DMSO : ≥ 50 mg/mL (143.49 mM)
SMILES O=C1COC2=CC(F)=CC=C2N1CCCN3CCC(CCCC)CC3
Molecular Weight (g/mol) 348.45
Peso formulazione 348.45
Percent Purity 99.25%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.