Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
- Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for stirring
- Raised display design protects electronics from spills
- Optional splash guard protection shield
- Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifica
Specifica
| Display | LED |
| Certificazione/Conformità | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| Tipo | PT1000 External probe 0-400C |