Learn More
Thermo Scientific™ Accessories for Digital Heating Cooling Drybath
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
No. of Places:
6
12
15
16
20
24
30
35
48
Dimensioni provetta:
0.5mL
1.5mL
10mm
12mm
13mm
15mL
15mm
16mm
18mm
20mm
50mL large
50mL small
Dimensione della confezione:
Pezzo
Descrizione
Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.
- Warranty: 2 years
Spezifikation
Spezifikation
| Capacità (sistema metrico) | 0.5 mL |
| Lighting | LED |
| No. of Places | 48 |
| Tipo | Block |
| Dimensioni provetta | 0.5mL |
| Dimensioni (L x P x A) | 3 x 3.9 x 1.6 in. (76.5 x 98 x 41 mm) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur