Learn More
Fisherbrand™ Accessories for Incubating Mini Shaker
Shop All Fisherbrand Products
Click to view available options
Product Type:
Centrifuge Tube Rack
Flask Clamp
Sticky Mat
Tube Rack with Adapter
Capacità:
1.5 - 2.0mL Tubes
100, 125 mL Flasks
15 mL Centrifuge Tubes
25 mL Flasks
250 mL Flasks
50 mL Centrifuge Tubes
50 mL Flasks
N/A
Unit Size:
Pezzo
Description
Accessories for incubating mini shaker
- Increase performance and flexibility
Specifications
Specifications
| Capacità | 25 mL Flasks |
| Product Type | Flask Clamp |
| Materiale | Stainless Steel |
| N. di posizioni | 20 |