Learn More
Thermo Scientific™ Acclaim™ Surfactant Plus HPLC Columns
Codice prodotto. 15210339 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
10 mm
100 mm
150 mm
250 mm
Diametro (sistema metrico):
2.1 mm
3 mm
4.6 mm
Dimensione particelle:
3 μm
5 μm
Dimensione della confezione:
Pezzo
pezzi 2
Codice prodotto. 15210339
Marca: Thermo Scientific™ 082773
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use for simultaneous separation of anionic, nonionic, cationic, and amphoteric surfactants with CAD and LC-MS detection.
For surfactant analysis using high-sensitivity detection, the column of choice is the Thermo Scientific™ Acclaim™ Surfactant Plus LC Columns. This new generation of high-efficiency silica-based columns features exceptionally low bleed for use with charged aerosol detectors (CAD) and mass spectrometers (MS). The columns provide selectivity similar to the Acclaim Surfactant columns for sensitive separation of a wide variety of surfactants. The advanced surface chemistry provides both reversed-phase and anion-exchange retention mechanisms and significantly improves the resolution of surfactants.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifica
Formato colonna Guard Cartridge
Diametro (sistema metrico) 4.6 mm
Lunghezza (sistema metrico) 10 mm
Pressione max 6000 psi (415 bar)
Dimensione particelle 5 μm
pH 2.5 to 7.5
Dimensione del poro 120 Å
Quantità Pack of 2
Surface Area 300 m²/g
Temperatura 50°C
Tipo USP L78
Fase stazionaria Proprietary
Product Line Acclaim
Materiale di impaccamento diossido di silicio ultrapuro completamente poroso, sferico
Tipo di colonna Specialty/Application Specific
Con tappo
Fase Reversed Phase and Ion Exchange
Tipo Guard Cartridge
Green Features Responsibly packaged