Learn More
Brand™ Dosatore per pipette Pro con batteria ibrida nichel-metallo
Sfoglia Tutto Brand Prodotti
Click to view available options
Colore:
Blu reale
Dark Blue
Magenta
Verde
Tipo di spina:
Europe (continent)
N/A - charger not included
Dimensione della confezione:
Pezzo
Descrizione
- One-handed delivery mode and selection
- Ergonomic design is perfectly balanced, weighing only 190g for fatigue-free pipetting even in prolonged operations.
- Adjustable motor speed limiter – a slower speed setting allows even more sensitive control of filling and dispensing rates for lower volume pipets.
- A flashing LED indicates 2 hours in advance when the battery needs charging. Approximately eight hours of continuous pipetting (with a 10mL pipet) without recharging.
- Direct outlet of liquid vapors protects against corrosion to ensure long instrument life.
- A 0.2μm hydrophobic membrane filter and a built-in check valve protect against liquid penetrating inside. Perfect fit for glass and plastic pipets from 0.1 to 200mL
Specifica
Specifica
| Tipo di batteria | All’idruro di nickel metallico |
| Colore | Magenta |
| Dimensione del poro | 0.2 μm |
| Tipo | Dosatore per pipette Pro |
| Tipo di filtro | Membrana idrofobica |
| Include | Nickel-metal hydride battery, 2 battery compartment covers, wall support, AC adapter (100-240V; 50/60Hz) and 2 spare membrane filters 0.2µm, sterile. 26304 without recharger |
| Tipo di spina | Europe (continent) |
| Quantità | 1 ciascuno |
| Peso (sistema metrico) | 190 g |
| Tensione | 100/240 V |
| Vedi altri risultati |