Learn More
Fisher Scientific™ Accu500 Water System
Codice prodotto. 15469699 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Tipo:
UV Ultrapure Water System
UV Ultrapure Water System w/ Dispenser
Ultrapure Water Purification System
Portata RO:
10 L/hr.
15 L/hr.
15 L/min.
20 L/hr.
30 L/hr.
Dimensione della confezione:
Pezzo
Codice prodotto. 15469699
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.
  • The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:
Ultrapure water
  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.
RO water
  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving
Specifica
Conducibilità <20μS/cm
Concentrazione di endotossine <0.001 EU/mL with a terminal ultrafiltration cartridge
Tipo di acqua prodotto Tap Water
Resistività 18.2 Mohm.cm
Tipo Ultrapure Water Purification System
Pressione acqua di alimentazione 1 to 6 bar
Modello Accu500
Particelle > 0,2 µM/mL <1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
Campo della portata dell'acqua purificata 1.5 L/min.
Portata RO 30 L/hr.
Temperature Range (Metric) 5°C to 45°C
Lunghezza (sistema metrico) 30 cm
Altezza (sistema metrico) 48 cm
Profondità (sistema metrico) 51 cm