Learn More
Thermo Scientific™ Accucore™ Polar Selectivity Kits
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
50 mm
100 mm
150 mm
Dimensione della confezione:
Pezzo
Descrizione
Use the Thermo Scientific™ Accucore™ Polar Selectivity Kits to verify optimum selectivity of Accucore HPLC columns for polar analytes.
Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution separations—without the elevated backpressures required by sub-2μm particles.
- Fast separations with excellent resolution and sensitivity
- Lower backpressures so that UHPLC is not required
- Kit contains 1 Accucore aQ HPLC Column, 1 Accucore PFP Column, and 1 Accucore HILIC Column
Specifica
Specifica
| Formato colonna | HPLC Column Kit |
| Diametro (sistema metrico) | 2.1 mm |
| Da utilizzare con | HPLC |
| Lunghezza (sistema metrico) | 100 mm |
| Forma particelle | Spherical Solid Core |
| Dimensione particelle | 2.6 μm |
| Dimensione del poro | 80 Å |
| Surface Area | 130 m²/g |
| Fase stazionaria | aQ C18, HILIC, PFP |
| Product Line | Accucore |
| Vedi altri risultati |