Learn More
Draeger Safety Accuro™ Gas Detector Pump
Codice prodotto. 10693781
Cambia vista
Click to view available options
Dimensioni (L x P x A):
205 x 240 x 60 mm
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P x A) Dimensione della confezione
10693781 205 x 240 x 60 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10693781 Fornitore Draeger Safety N. del fornitore 6400260
  • ONLINE EXCLUSIVE PRICE
    1959.60€
    1450.03€ / pezzi 1
    Risparmia 509.57€
    26% Sconto
Spedizione stimata: 09-12-2026
Draeger Safety Accuro™ Gas Detector Pump is a manually operated bellows pump that draws a calibrated 100 mL sample through the short-term Draeger-Tubes.
  • One-Hand Operation
  • Very Low Maintenance
  • End-of-Stroke Indicator
  • Built-in automatic Stroke Counter
Specifica
Dimensioni (L x P x A) 205 x 240 x 60 mm
Peso (sistema metrico) 0.92 kg