missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acetyl-CoA Carboxylase Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA577844
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Pig (94%), Chicken (83%), Sheep (94%), Rhesus Monkey (98%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.
Specifica
| Acetyl-CoA Carboxylase | |
| Polyclonal | |
| Unconjugated | |
| Acaca | |
| A1Z784_DROME; A530025K05Rik; Acac; Acaca; ACACAD; Acc; ACC1; ACCA; ACC-alpha; ACC-PA; ACC-PB; ACC-PC; ACC-PE; ACC-PF; ACC-PG; aceto-acetyl-CoA-thiolase; acetyl CoA carboxylase; acetyl coenzyme A carboxylase; Acetyl-CoA carboxylase; Acetyl-CoA carboxylase 1; acetyl-CoA carboxylase 265; acetyl-CoA carboxylase alpha; acetyl-CoA-carboxylase; acetyl-coenzyme A carboxylase; acetyl-coenzyme A carboxylase alpha; ACoT; Biotin carboxylase; CG11198; CG11198-PA; CG11198-PB; CG11198-PC; CG11198-PE; CG11198-PF; CG11198-PG; CG8723; dACC; DmACC; Dmel\CG11198; Dmel_CG11198; EC 6.3.4.14; EC 6.4.1.2; FBgn0043811; GH12002p; Gm738; I79_006999; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 104371, 107476, 31, 60581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.55 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P11497, Q13085, Q5SWU9 | |
| Acaca, ACC | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Acetyl-CoA Carboxylase 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto