Learn More
WTW™ Acid Capacity Cell and Reagent Test Kits
Click to view available options
N. di test:
120
Dimensione della confezione:
Pezzo
Descrizione
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Photometers |
| Include | Certificates |
| N. di test | 120 |
| Range | KS4.3 0.40 - 8.00mmol/L; 20 - 400 mg/L CaCO3 |
| Sufficiente per | 120 Tests |
| Metodo test | Photometric |
| Tipo | Acid Capacity cell/reagent test kit |