missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT11 Antibody (J4B2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-04289R
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ACOT11 Monoclonal specifically detects ACOT11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| ACOT11 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACOT11 | |
| Recombinant human ACOT11 (19-250aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 26027 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| J4B2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Acyl-CoA thioester hydrolase 11, acyl-CoA thioesterase 11BFIT1, Adipose-associated thioesterase, BFIT2, BFITDKFZp667O1916, Brown fat-inducible thioesterase, EC 3.1.2.-, EC 3.1.2.1, KIAA0707acyl-coenzyme A thioesterase 11, STARD14, START domain containing 14, THEAStAR-related lipid transfer (START) domain containing 14, THEM1, thioesterase superfamily member 1, thioesterase, adipose associated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human ACOT11 mAb, clone J4B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ACOT11 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto