Learn More
Cytiva Acro™ 50 Vent Devices with HDC II Media
Click to view available options
:
6 μm
Descrizione
- 0.2 micron rated and contain a hydrophobic PTFE membrane
- The large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
Specifica
Specifica
| Dimensione del poro | 6 μm |
| Sterilità | Non-sterile |
| Quantità | 18/Pk. |