Learn More
Brady™ Acrylic Retro Reflective Sheeting Marking Tape
Click to view available options
Colore:
Black and White Striped
Black and Yellow Striped
Red on White
Dimensioni (L x P):
4.57 m x 50.80 mm
4.57 m x 76.20 mm
Dimensione della confezione:
1 ROLL
Descrizione
- The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse.
- These tapes give an extra margin of safety during blackouts, at night, or in low-light conditions.
Specifica
Specifica
| Alternate Part Number | B957 3 in. X5YDS R/W STRIPED REFLECTIVE TAPE |
| Certificazione/Conformità | ASTM D 2979, ASTM D 523 |
| Dimensioni (L x P) | 4.57 m x 76.20 mm |
| Finitura | Semi-gloss |
| Da utilizzare con (applicazione) | Utility plant markings, Reflective Bradystake Warning Stakes, Reflective warning stripes |
| Lunghezza (sistema imperiale) | 15 ft. |
| Lunghezza (sistema metrico) | 4.57 m |
| Quantità confezione | 1 Roll |
| Intervallo di temperatura (sistema imperiale) | -40°F to +158°F |
| Temperature Range (Metric) | -40°C to +70°C |
| Vedi altri risultati |