Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Click to view available options
Volume (sistema metrico):
1 μL
5 μL
10 μL
15 μL
20 μL
25 μL
40 μL
50 μL
60 μL
70 μL
75 μL
80 μL
90 μL
100 μL
120 μL
150 μL
200 μL
250 μL
300 μL
400 μL
500 μL
600 μL
700 μL
750 μL
800 μL
900 μL
1000 μL
Dimensione della confezione:
Pezzo
Descrizione
- Air-displacement fixed-volume pipette.
- Exceptional, user-friendly ergonomics.
- Justip™ adjustable tip ejector fitting most tips.
- Shock, UV-light and autoclaving resistance.
- Ultra-soft plunger stroke.
- Available in a variety of volume models.
Specifica
Specifica
| Accuracy | <±0.7% |
| Certificazione/Conformità | ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices). |
| Autoclavabile | Autoclavable |
| Colore | Giallo |
| Modello | 815 |
| Tipo | Micropipette |
| Volume (sistema metrico) | 50 μL |
| Precisione | <0.5% |
| Quantità | 1 Each |
| Warranty | 36 Months |