missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584955
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: LPHLIYRTPL LPDPLGCREP GCLFAVPAQS APPNRPRLRR KRQVRRGHPT VHSETKYVEL IVINDHQLFE QMR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 93%.
Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
Specifica
| ADAM11 | |
| Polyclonal | |
| Unconjugated | |
| ADAM11 | |
| a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4185 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75078 | |
| ADAM11 | |
| Recombinant protein corresponding to Human ADAM11. Recombinant protein control fragment (Product #RP-108268). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto