Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
Sfoglia Tutto Eppendorf Prodotti
Click to view available options
Capacità:
1 x 1 L Nalgene Bottle
1 x 500 mL Conical Bottle
1 x 750 mL Flat-bottom Bottle
16 x 5 mL Microtubes, 16 x 15 mL Conical Tubes, 1 MTP/DWP
21 x 9 mL Round Bottom Tubes
26 x 7.5 to 12 mL Round Bottom Tubes
30 x 4 to 8 mL Round Bottom Tubes
46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS)
5 x MTP/DWP/PCR Plates
7 x 50 mL Conical Tubes, 1 x 96/384-well Microplate, 1 x 250 mL Flat-bottom Bottle, 1 x 175 to 225 mL Conical Bottle
7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes
9 x 50 mL Conical Tubes, 1 Microplate
Tipo di provetta:
Conical
Flat Bottom
Flat/Conical Bottom
Microtube, Conical
PCR
Round Bottom
Round Bottom (FACS)
Skirted Bottom, Oak Ridge
No. of Places:
1
16/16/1
21
26
30
46
5
7/1/1/1
7/7
9/1
Dimensione della confezione:
pezzi 2
Descrizione
- Suitable for S-4xUniversal rotor buckets
- Accommodates selected array of tubes, bottles, or plates
Specifica
Specifica
| Tipo di provetta | Round Bottom (FACS) |
| Capacità | 46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS) |
| Da utilizzare con (apparecchiatura) | S-4xUniversal Rotor |
| No. of Places | 46 |
| N. per confezione | 2 |