Learn More
Techne™ Additional Loading Dock
Click to view available options
:
2 Centrifuge Adapters
Descrizione
- This ergonomic plate loading dock allows for easy loading of a plate
- LED Backlight allows the user to easily visualise sample loading
- Plate adapter fits standard centrifuge rotors
Specifica
Specifica
| Content And Storage | 2 Centrifuge Adapters |
| Da utilizzare con (apparecchiatura) | Prime Pro 48 Real-Time PCR System |
| Product Type | Additional Loading Dock |