missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538845
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase [provided by RefSeq, Jul 2008].
Specifica
| Adenylate Cyclase 8 | |
| Polyclonal | |
| Unconjugated | |
| ADCY8 | |
| AarF domain containing kinase 9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114, 11514, 29241 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40146, P40145, P97490 | |
| ADCY8 | |
| A synthetic peptide derived from the internal region of human ADCY8 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto