missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipophilin / Perilipin-2 Mouse, Clone: ADFP/1365, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 123-MSM1-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Adipophilin / Perilipin-2 | |
Cytoplasmic | |
ADFP/1365 | |
Unconjugated | |
XP_016869748 | |
Mouse | |
48kDa | |
Protein A/G Purified | |
RUO | |
123 | |
IgG1 κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
PLIN2 | |
Recombinant human Adipophilin protein fragment (aa249-376) | |
HepG2 or JAR cells. Liver, Adrenal gland or Cerebellum. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |