Learn More
Tocris Bioscience™ AEBSF
Click to view available options
:
100mg
Descrizione
AEBSF is a broad spectrum, irreversible inhibitor of serine proteases. Also inhibits NADPH oxidase activation in vitro. Attenuates airway inflammation in a mouse model of airway allergy.
Specifica
Specifica
| Quantità | 100 mg |
| Product Type | AEBSF |
| Purezza | >98% |
| Peso molecolare | 239.69 |
| Inibitori | AEBSF |
For Research Use Only