Learn More
Agarose gel loading dye (6X), glycerol based, bromophenol free, Thermo Scientific™
Click to view available options
Quantità:
25 mL
50 mL
Dimensione della confezione:
25mL
50mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Agarose gel loading dye |
| Nota nome | glycerol based, bromophenol free |
| Forma fisica | Liquid |
| Concentrazione | 6X |
| Quantità | 25 mL |
| Numero MDL | MFCD00675058 |