Learn More
AIM2 Mouse anti-Human, Unconjugated, Clone: 3E12A8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66902-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The AIM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AIM2. This antibody recognizes human, mouse, rat, rabbit antigen. The AIM2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
| AIM2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit |
| Western Blot, Co-immunoprecipitation (Co-IP) | |
| 3E12A8 | |
| Unconjugated | |
| 20 μL | |
| 9447 | |
| IgG1 |
Titolo del prodotto
Individuate un'opportunità di miglioramento?Dela en innehållskorrigering