Learn More
Abeba™ Air-Cushion 32370 Shoes
Click to view available options
Size:
35
36
38
39
41
42
43
44
45
46
47
Unit Size:
1 paio
Description
- CE EN ISO 20347:2012 O1 SRA
- Microfiber
- White
- Slip resistance PU outsole sole
- Integrated footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Instep area with elastic band
- Decontaminable
Specifications
Specifications
| Certificazione/Conformità | CE EN ISO 20347:2012 O1 SRA |
| Colore | White |
| Materiale | Microfiber |
| Size | 35 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 32370 |