Learn More
Abeba™ Air-Cushion Sole 2600 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 O2 SRA
- Grained leather
- White
- Slip resistance PU outsole sole
- Replaceable insole (article no. 3551)
- Sanitized treated
- Antistatic
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 O2 SRA |
| Colore | White |
| Materiale rivestimento interno | Microfiber |
| Materiale | Leather |
| Size | 42 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 2600 |