Learn More
AKAP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12591-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody can recognize all the isoforms of AKAP7.
This gene encodes a member of the A-kinase anchoring protein family, a group of functionally related proteins that bind to a regulatory subunit of cAMP-dependent protein kinase A and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.Specifica
| AKAP7 | |
| Polyclonal | |
| Unconjugated | |
| AKAP7 | |
| A kinase anchor protein 18 kDa, AKAP 18, AKAP 7 isoforms alpha and beta, AKAP18, AKAP7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361458, 432442, 9465 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43687, O55074, Q6JP77 | |
| Akap7 | |
| AKAP7 Fusion Protein Ag3297 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |