Learn More
Al-23 Grinding Pieces, H: 10mm, 440 Pieces/Kg, Thermo Scientific Chemicals
Product Code. 15431307
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431307 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15431307 Supplier Thermo Scientific Alfa Aesar Supplier No. 033190.A1
 Restricted Item
Spedizione stimata: 15-12-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Grinding Pieces
Quantity 1 kg
Form Grinding Pieces
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.