Learn More
Al-23 Multibore Insulating Tube, O. D.: 8mm, Two 1.6mm Bores, Thermo Scientific™
Click to view available options
Quantità:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Dimensione della confezione:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Al-23 Multibore Insulating Tube |
| Nota nome | Two bores |
| Quantità | 560 mm |