Learn More
Al-23 Multibore Round Tube, O.D.: 4mm, Four 0.8mm Bores, Thermo Scientific™
Click to view available options
Quantità:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Dimensione della confezione:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Al-23 Multibore Round Tube |
| Nota nome | Four bores |
| Quantità | 250 mm |