Learn More
Al-23 Tube (both ends open);OD (mm), 35;ID (mm), 29
Click to view available options
Quantità:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Dimensione della confezione:
1000mm
100mm
200mm
300mm
600mm
800mm
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale o nome chimico | Al-23 Tube |
| Nota nome | Both ends open |
| Quantità | 100 mm |