Learn More
Thermo Scientific Chemicals Al-25 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 160 x 145 x 125;Vol (ml), 1760
Click to view available options
Capacità (sistema metrico):
1760 mL
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Diametro (sistema metrico) interno | 16cm |
| Materiale | Al-25 |
| Quantità | 1 Piece |
| Capacità (sistema metrico) | 1760 mL |
| Diametro (sistema metrico) esterno | 12.5cm |
| Altezza (sistema metrico) | 14.5cm |
| Product Type | Cylindrical Al-25 Crucibles |