Learn More
VEE GEE Alcohol Hydrometer
Codice prodotto. p-7250815
Click to view available options
Alcohol Range:
0% to 20%
105% to 125%
125% to 145%
145% to 165%
165% to 185%
185 to 206%
20 to 40%
40 to 60%
60 to 80%
75% to 95%
80 to 120%
90 to 110%
Incrementi:
0.2%/±0.2%
0.2%/±0.3%
0.2%/±0.4%
Dimensione della confezione:
Pezzo
Codice prodotto. 17866523
Marca: VEE GEE 6613Q
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
  • Alcohol proof range differ depending on model
  • IRS Specification Size differ depending on model
  • li>Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifica
Tipo Alcohol Hydrometer
Incrementi 0.2%/±0.2%
Lunghezza (sistema metrico) 30.5 cm
Descrizione Alcohol Hydrometer, Proof Scale, IRS Specification, Size Q
Materiale Glass
Temperature of Standardization 15.5°C (60°F)
Alcohol Range 165% to 185%