Learn More
VEE GEE Alcohol Hydrometer with Certification
Click to view available options
Alcohol Range:
0 to 20%
105 to 125%
125 to 145%
145 to 165%
165 to 185%
185 to 206%
20 to 40%
40 to 60%
60 to 80%
75 to 95%
80 to 120%
90 to 110%
Incrementi:
0.20%
0.50%
Dimensione della confezione:
Pezzo
Descrizione
- Alcohol proof range differ depending on model
- IRS Specification Size differ depending on model li>Calibration for use at 15.5°C/60°F
- Approx. Length: 305 mm
Specifica
Specifica
| Tipo | Alcohol Hydrometer |
| Incrementi | 0.20% |
| Lunghezza (sistema metrico) | 30.5 cm |
| Descrizione | Alcohol Hydrometer, Proof Scale, IRS Specification, Size H, with Certification |
| Temperature of Standardization | 15.5°C (60°F) |
| Alcohol Range | 40 to 60% |