Learn More
ALDH1A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25167-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.Specifica
| ALDH1A3 | |
| Polyclonal | |
| Unconjugated | |
| ALDH1A3 | |
| Aldehyde dehydrogenase 6, ALDH1A3, ALDH1A6, ALDH6, RALDH 3, RALDH3, Retinaldehyde dehydrogenase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 220, 56847 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47895, Q9JHW9 | |
| Aldh1a3 | |
| ALDH1A3 Fusion Protein Ag18605 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |