Learn More
Alloy Steel, Ni/Cr/Mo, AISI 4340, G43400, 100g, ARMI∣MBH
Codice prodotto. 17973516
Click to view available options
Quantità:
100 g
Dimensione della confezione:
100g
Codice prodotto. 17973516
Marca: ARMI/MBH IARM31GC
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Alloy Steel, Ni/Cr/Mo, AISI 4340, G43400, 100g
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Boron/Zirconium/Titanium/Niobium/Vanadium/Arsenic/Tungsten/Nitrogen/Tin/Cobalt/Phosphorus/Aluminium/Copper/Molybdenum/Silicon/Carbon/Manganese/Chromium/Nickel
CAS 7439-89-6
Quantità 100 g
Forma Pieces
Concentration or Composition (by Analyte or Components) Boron 0.0004 %/Zirconium 0.0012 %/Titanium 0.0015 %/Niobium 0.0023 %/Vanadium 0.0036 %/Arsenic 0.004 %/Tungsten 0.004 %/Nitrogen 0.0069 %/Tin 0.0077 %/Cobalt 0.0083 %/Phosphorus 0.0136 %/Aluminium 0.0214 %/Copper 0.183 %/Molybdenum 0.223 %/Silicon 0.262 %/Carbon 0.4 %/Manganese 0.689 %/Chromium 0.82 %/Nickel 1.814 %