missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-1-Antitrypsin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16382-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.Specifica
| Alpha-1-Antitrypsin | |
| Polyclonal | |
| Unconjugated | |
| SERPINA1 | |
| A1A, A1AT, AAT, Alpha 1 antiproteinase, Alpha 1 antitrypsin, Alpha 1 protease inhibitor, PI, PI1, PRO2209, PRO2275, Serpin A1, SERPINA1, SPAAT, alpha1-antitrypsin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20700, 24648, 5265 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01009, P07758, P17475 | |
| SERPINA1, Serpina1a | |
| Alpha-1-Antitrypsin Fusion Protein Ag9369 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto