missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody (1A4/asm-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
638.00€ - 837.00€
Specifica
| Antigene | alpha-Smooth Muscle Actin |
|---|---|
| Clone | 1A4/asm-1 |
| Concentrazione | 1.0 mg/mL |
| Diluizione | Western Blot, Simple Western, Flow Cytometry, ELISA 1-5ug/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/mL, Immunohistochemistry-Frozen 0.5-1ug/mL, Flow (Intracellular), CyTOF-ready |
| Applicazioni | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18433622
|
Novus Biologicals
NBP2-34522-0.2mg |
0.2 mg |
837.00€
0.02mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18426671
|
Novus Biologicals
NBP2-34522-0.1mg |
0.1 mg |
638.00€
0.10mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOFSpecifica
| alpha-Smooth Muscle Actin | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 59 | |
| IgG2a κ | |
| Protein A purified |
| 1A4/asm-1 | |
| Western Blot, Simple Western, Flow Cytometry, ELISA 1-5ug/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/mL, Immunohistochemistry-Frozen 0.5-1ug/mL, Flow (Intracellular), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit, Sheep | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| Alpha-Smooth Muscle Actin Antibody (1A4/asm-1) is made to the N-Terminal decapeptide of Alpha smooth muscle isoform of actin and conjugated to KLH. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto