Learn More
alpha-Smooth Muscle Actin Mouse anti-Human, Alexa Fluor 350, Clone: 1A4, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems IC1420U-100UG
Descrizione
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifica
| alpha-Smooth Muscle Actin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human alpha-Smooth Muscle Actin. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 1A4 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACTA2 | |
| Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha-smooth muscle actin (1) | |
| RUO | |
| 59 | |
| Human | |
| IgG2a |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
For Research Use Only (RUO)