missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein, p Ser129 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
242.00€ - 822.00€
Specifica
| Antigene | alpha-Synuclein (p Ser129) |
|---|---|
| Concentrazione | 0.5 mg/ml |
| Diluizione | Western Blot 1:1000, ELISA, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 |
| Classificazione | Monoclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18225375
|
Novus Biologicals
NBP2-61121 |
100 μL |
822.00€
100µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18612787
|
Novus Biologicals
NBP2-61121-0.025ml |
0.025 mL |
242.00€
0.03mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
alpha-Synuclein Polyclonal specifically detects alpha-Synuclein in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifica
| alpha-Synuclein (p Ser129) | |
| Western Blot 1:1000, ELISA, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 | |
| Unconjugated | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| This alpha-Synuclein [p Ser129] Antibody was developed against Synthetic peptide of human alpha-Synuclein pSer129 (40-140 aa), conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| This alpha-Synuclein [p Ser129] Antibody is specific for phosphorylated serine 129 on alpha-synuclein. Does not detect unphosphorylated serine 129 alpha-synuclein. Detects bands at 100, 75, 45, 15 kDa. Bands above 15 kDa are oligomers. |
| 0.5 mg/ml | |
| Monoclonal | |
| Rabbit | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| SNCA | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title